Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fibpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212861.2 | NP_998026.1 | 4 - 5 | 621 | 67 | |
XM_009295296.3 | XP_009293571.1 | 4 - 5 | 742 | 67 | ||
XM_009295297.3 | XP_009293572.1 | 4 - 5 | 517 | 67 | ||
XM_021468797.1 | XP_021324472.1 | 4 - 5 | 742 | 67 | ||
XM_021468798.1 | XP_021324473.1 | 4 - 5 | 742 | 67 | ||
GenBank mRNA | AY427781.1 | - | 3 - 4 | 417 | 67 | |
BC076477.1 | - | 4 - 5 | 666 | 67 | ||
BC085535.1 | - | 4 - 5 | 621 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |