Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ciapin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003738.1 | NP_001003738.1 | 3 - 4 | 343 | 88 | |
XM_005174319.4 | XP_005174376.3 | 3 - 4 | 448 | 88 | ||
XM_005174320.4 | XP_005174377.2 | 3 - 4 | 412 | 88 | ||
XM_005174321.4 | XP_005174378.1 | 3 - 4 | 414 | 88 | ||
GenBank mRNA | BC078404.1 | - | 3 - 4 | 343 | 88 | |
BC164092.1 | - | 2 - 3 | 336 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |