Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps4x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005589.1 | NP_001005589.1 | 4 - 5 | 542 | 135 | |
GenBank mRNA | BC081584.1 | - | 4 - 5 | 542 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |