Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: samsn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006039.2 | NP_001006039.2 | 7 - 8 | 972 | 88 | |
XM_005169164.4 | XP_005169221.1 | 15 - 16 | 1922 | 88 | ||
XM_009305561.3 | XP_009303836.1 | 15 - 16 | 1919 | 88 | ||
GenBank mRNA | BC083374.1 | - | 7 - 8 | 945 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |