Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pygma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020628.1 | NP_001018464.1 | 10 - 11 | 1307 | 85 | |
GenBank mRNA | BC095379.1 | - | 10 - 11 | 1307 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |