Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clvs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025545.2 | NP_001020716.1 | 2 - 3 | 995 | 89 | |
GenBank mRNA | BC116475.1 | - | 2 - 3 | 995 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |