Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtmr7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030126.1 | NP_001025297.1 | 8 - 9 | 1193 | 80 | |
XM_005168352.4 | XP_005168409.1 | 8 - 9 | 1221 | 80 | ||
GenBank mRNA | BC097161.1 | - | 8 - 9 | 1193 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |