Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eef1db | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030147.2 | NP_001025318.1 | 3 - 4 | 277 | 73 | |
XM_005160919.4 | XP_005160976.1 | 5 - 6 | 1166 | 73 | ||
GenBank mRNA | BC081476.1 | - | 3 - 4 | 259 | 73 | |
BC097116.1 | - | 3 - 4 | 259 | 73 | ||
EH506100.1 | - | 3 - 4 | 288 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |