Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dync1h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042745.1 | NP_001036210.1 | 13749 | 94 | ||
GenBank mRNA | DQ323903.1 | - | 13749 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio |