Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plekho1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110463.1 | NP_001103933.1 | 3 - 4 | 423 | 146 | |
XM_005159748.4 | XP_005159805.1 | 3 - 4 | 674 | 146 | ||
GenBank mRNA | AB303429.1 | - | 1 - 2 | 113 | 146 | |
BC134933.1 | - | 3 - 4 | 423 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |