Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myo7aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152983.1 | NP_694515.1 | 20 - 21 | 2528 | 73 | |
XM_021467573.1 | XP_021323248.1 | 21 - 22 | 2679 | 73 | ||
GenBank mRNA | AJ404001.1 | - | 20 - 21 | 2528 | 73 | |
AJ404002.1 | - | 19 - 20 | 2408 | 73 | ||
BC163570.1 | - | 19 - 20 | 2416 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |