Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mpp5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194363.1 | NP_919344.1 | 3 - 4 | 784 | 75 | |
XM_009293174.3 | XP_009291449.1 | 4 - 5 | 886 | 75 | ||
GenBank mRNA | AF510111.1 | - | 3 - 4 | 784 | 75 | |
BC163251.1 | - | 1 - 2 | 518 | 75 | ||
BC163277.1 | - | 1 - 2 | 518 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |