Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mpp5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194363.1 | NP_919344.1 | 8 - 9 | 1465 | 98 | |
XM_009293174.3 | XP_009291449.1 | 9 - 10 | 1567 | 98 | ||
GenBank mRNA | AF510111.1 | - | 8 - 9 | 1465 | 98 | |
BC163251.1 | - | 6 - 7 | 1199 | 98 | ||
BC163277.1 | - | 6 - 7 | 1199 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |