Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pkma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199333.1 | NP_955365.1 | 5 - 6 | 621 | 104 | |
XM_009293364.2 | XP_009291639.1 | 5 - 6 | 734 | 104 | ||
GenBank mRNA | BC045421.1 | - | 5 - 6 | 621 | 104 | |
BC067143.1 | - | 4 - 5 | 104 | |||
BC164692.1 | - | 4 - 5 | 600 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |