Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: serpinb1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199936.1 | NP_956230.1 | 4 - 5 | 518 | 105 | |
XM_005174205.4 | XP_005174262.1 | 4 - 5 | 710 | 105 | ||
GenBank mRNA | BC053300.1 | - | 4 - 5 | 518 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |