Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: serpinb1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199936.1 | NP_956230.1 | 5 - 6 | 660 | 71 | |
XM_005174205.4 | XP_005174262.1 | 5 - 6 | 852 | 71 | ||
GenBank mRNA | BC053300.1 | - | 5 - 6 | 660 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |