Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: laptm4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200098.2 | NP_956392.1 | 6 - 7 | 790 | 76 | |
GenBank mRNA | BC059484.1 | - | 6 - 7 | 719 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |