Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201013.3 | NP_957307.3 | 6 - 7 | 754 | 86 | |
XM_005161962.4 | XP_005162019.1 | 6 - 7 | 770 | 86 | ||
XM_021469854.1 | XP_021325529.1 | 6 - 7 | 767 | 86 | ||
GenBank mRNA | BC066391.1 | - | 6 - 7 | 733 | 86 | |
BC154825.1 | - | 6 - 7 | 696 | 86 | ||
EH561147.1 | - | 4 - 5 | 760 | 86 | ||
EH594482.1 | - | 6 - 7 | 781 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |