Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201013.3 | NP_957307.3 | 7 - 8 | 1019 | 77 | |
XM_005161962.4 | XP_005162019.1 | 7 - 8 | 1035 | 77 | ||
XM_021469854.1 | XP_021325529.1 | 7 - 8 | 1032 | 77 | ||
GenBank mRNA | BC045915.1 | - | 6 - 7 | 825 | 77 | |
BC066391.1 | - | 7 - 8 | 998 | 77 | ||
BC154825.1 | - | 7 - 8 | 961 | 77 | ||
EH561147.1 | - | 3 - 4 | 495 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |