Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ivns1abpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201483.1 | NP_958891.1 | 9 - 10 | 1180 | 83 | |
XM_021479057.1 | XP_021334732.1 | 3 - 4 | 638 | 83 | ||
GenBank mRNA | BC045449.1 | - | 9 - 10 | 1180 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |