Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glud1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212576.1 | NP_997741.1 | 10 - 11 | 1479 | 91 | |
GenBank mRNA | BC066370.1 | - | 10 - 11 | 1479 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |