Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc20a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212588.1 | NP_997753.1 | 5 - 6 | 813 | 63 | |
XM_005155625.4 | XP_005155682.1 | 6 - 7 | 1493 | 63 | ||
GenBank mRNA | BC057497.1 | - | 5 - 6 | 813 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |