Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tead1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212847.2 | NP_998012.1 | 4 - 5 | 421 | 72 | |
XM_005174314.3 | XP_005174371.1 | 3 - 4 | 536 | 72 | ||
XM_009297895.3 | XP_009296170.1 | 2 - 3 | 434 | 72 | ||
GenBank mRNA | BC055211.1 | - | 4 - 5 | 422 | 72 | |
BC097055.1 | - | 3 - 4 | 486 | 72 | ||
BC164522.1 | - | 1 - 2 | 233 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |