Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bzw1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213092.1 | NP_998257.1 | 11 - 12 | 1294 | 97 | |
XM_005167390.4 | XP_005167447.1 | 11 - 12 | 1384 | 97 | ||
GenBank mRNA | BC044401.1 | - | 11 - 12 | 1314 | 97 | |
BC066527.1 | - | 11 - 12 | 1294 | 97 | ||
BC091971.1 | - | 11 - 12 | 1316 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |