Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002556.1 | NP_001002556.1 | 4 - 5 | 451 | 67 | |
GenBank mRNA | BC076246.1 | - | 4 - 5 | 451 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |