Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdf2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003730.1 | NP_001003730.2 | 2 - 3 | 389 | 80 | |
GenBank mRNA | BC078401.1 | - | 2 - 3 | 389 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |