Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003869.1 | NP_001003869.1 | 3 - 4 | 824 | 122 | |
GenBank mRNA | AY648791.1 | - | 3 - 4 | 824 | 122 | |
BC093104.1 | - | 3 - 4 | 772 | 122 | ||
BC115114.1 | - | 3 - 4 | 122 | |||
BC121728.1 | - | 3 - 4 | 122 | |||
BC129281.1 | - | 3 - 4 | 122 | |||
BC134116.1 | - | 3 - 4 | 122 | |||
BC164482.1 | - | 2 - 3 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |