Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: st3gal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004012.1 | NP_001004012.1 | 2 - 3 | 610 | 67 | |
XM_009293548.3 | XP_009291823.1 | 3 - 4 | 2375 | 67 | ||
XM_009293550.3 | XP_009291825.1 | 3 - 4 | 2134 | 67 | ||
GenBank mRNA | AJ705062.2 | - | 2 - 3 | 610 | 67 | |
AJ783741.1 | - | 2 - 3 | 610 | 67 | ||
BC162919.1 | - | 2 - 3 | 659 | 67 | ||
BC162930.1 | - | 2 - 3 | 659 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |