Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: polr3gla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004639.1 | NP_001004639.2 | 3 - 4 | 363 | 87 | |
GenBank mRNA | BC081380.1 | - | 3 - 4 | 363 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |