Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: polr3gla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004639.1 | NP_001004639.2 | 6 - 7 | 562 | 199 | |
GenBank mRNA | BC081380.1 | - | 6 - 7 | 562 | 199 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |