Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcdh15a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012482.1 | NP_001012500.1 | 16 - 17 | 2395 | 79 | |
XM_009307525.2 | XP_009305800.1 | 16 - 17 | 2407 | 79 | ||
XM_009307527.2 | XP_009305802.1 | 15 - 16 | 2392 | 79 | ||
XM_017358709.2 | XP_017214198.1 | 16 - 17 | 2407 | 79 | ||
XM_017358710.2 | XP_017214199.1 | 16 - 17 | 2407 | 79 | ||
XM_017358711.2 | XP_017214200.1 | 16 - 17 | 2407 | 79 | ||
XM_017358712.2 | XP_017214201.1 | 16 - 17 | 2407 | 79 | ||
XM_017358713.2 | XP_017214202.1 | 16 - 17 | 2407 | 79 | ||
XM_017358714.2 | XP_017214203.1 | 16 - 17 | 2407 | 79 | ||
GenBank mRNA | AY772390.1 | - | 16 - 17 | 2395 | 79 | |
KY432405.1 | - | 16 - 17 | 2394 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |