Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atad1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024421.2 | NP_001019592.1 | 2 - 3 | 343 | 126 | |
GenBank mRNA | BC095151.1 | - | 2 - 3 | 344 | 126 | |
BC164188.1 | - | 1 - 2 | 209 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |