Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tas1r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039628.1 | NP_001034717.1 | 6 - 7 | 1618 | 66 | |
GenBank mRNA | AB200902.1 | - | 6 - 7 | 1618 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Danio rerio Probe spans exons |