Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt5c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045386.1 | NP_001038851.1 | 1 - 2 | 56 | 79 | |
GenBank mRNA | BC122216.1 | - | 1 - 2 | 56 | 79 | |
BC164669.1 | - | 1 - 2 | 90 | 79 | ||
GU299162.1 | - | 1 - 2 | 29 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |