Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131447.1 | NP_571522.1 | 6 - 7 | 1926 | 65 | |
GenBank mRNA | AF162696.1 | - | 6 - 7 | 1926 | 65 | |
AJ245966.1 | - | 6 - 7 | 1535 | 65 | ||
BC162094.1 | - | 6 - 7 | 1604 | 65 | ||
BC162106.1 | - | 6 - 7 | 1604 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |