Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131708.2 | NP_571783.2 | 3 - 4 | 484 | 88 | |
GenBank mRNA | AF541952.1 | - | 3 - 4 | 467 | 88 | |
AJ297822.1 | - | 1 - 2 | 289 | 88 | ||
BC092664.1 | - | 3 - 4 | 481 | 88 | ||
BC165931.1 | - | 3 - 4 | 494 | 88 | ||
EH490900.1 | - | 3 - 4 | 514 | 88 | ||
EH517238.1 | - | 2 - 3 | 393 | 88 | ||
JQ999998.1 | - | 3 - 4 | 478 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |