Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131756.1 | NP_571831.1 | 6 - 7 | 765 | 83 | |
GenBank mRNA | AJ009864.1 | - | 6 - 7 | 765 | 83 | |
BC071295.1 | - | 6 - 7 | 740 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |