Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zdhhc8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178304.3 | NP_840089.2 | 9 - 10 | 1418 | 97 | |
GenBank mRNA | AB085761.1 | - | 9 - 10 | 1110 | 97 | |
AY871211.1 | - | 9 - 10 | 1418 | 97 | ||
BC049439.1 | - | 9 - 10 | 1212 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |