Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ilk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200571.1 | NP_956865.1 | 6 - 7 | 770 | 102 | |
GenBank mRNA | BC056593.1 | - | 6 - 7 | 770 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |