Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arpc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201343.2 | NP_958500.1 | 6 - 7 | 905 | 124 | |
GenBank mRNA | AY398313.1 | - | 6 - 7 | 903 | 124 | |
BC048036.1 | - | 6 - 7 | 896 | 124 | ||
BC164199.1 | - | 5 - 6 | 742 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |