Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201458.2 | NP_958866.1 | 12 - 13 | 1479 | 65 | |
GenBank mRNA | BC046072.1 | - | 12 - 13 | 1480 | 65 | |
BC063234.1 | - | 12 - 13 | 1440 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |