Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201458.2 | NP_958866.1 | 2 - 3 | 198 | 77 | |
GenBank mRNA | BC046072.1 | - | 2 - 3 | 199 | 77 | |
BC063234.1 | - | 2 - 3 | 159 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |