Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201458.2 | NP_958866.1 | 8 - 9 | 891 | 67 | |
GenBank mRNA | BC046072.1 | - | 8 - 9 | 892 | 67 | |
BC063234.1 | - | 8 - 9 | 852 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |