Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grcc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212646.1 | NP_997811.1 | 2 - 3 | 159 | 65 | |
GenBank mRNA | AY398355.1 | - | 2 - 3 | 159 | 65 | |
BC071485.1 | - | 2 - 3 | 144 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |