Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nlk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212956.1 | NP_998121.1 | 8 - 9 | 1210 | 73 | |
GenBank mRNA | AY562552.1 | - | 8 - 9 | 1210 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |