Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mfsd14a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213527.2 | NP_998692.2 | 6 - 7 | 889 | 99 | |
GenBank mRNA | BC045310.1 | - | 6 - 7 | 887 | 99 | |
BC097075.1 | - | 6 - 7 | 883 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |