Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002666.1 | NP_001002666.2 | 7 - 8 | 903 | 71 | |
XM_021477460.1 | XP_021333135.1 | 7 - 8 | 1042 | 71 | ||
GenBank mRNA | BC076553.1 | - | 7 - 8 | 903 | 71 | |
BC163973.1 | - | 7 - 8 | 817 | 71 | ||
HM138690.1 | - | 7 - 8 | 789 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |