Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcgf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007158.2 | NP_001007159.1 | 6 - 7 | 624 | 71 | |
GenBank mRNA | BC134012.1 | - | 6 - 7 | 624 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |