Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtmr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014330.2 | NP_001014352.1 | 4 - 5 | 554 | 74 | |
XM_005161411.4 | XP_005161468.1 | 3 - 4 | 556 | 74 | ||
GenBank mRNA | BC091956.1 | - | 4 - 5 | 558 | 74 | |
BC165859.1 | - | 4 - 5 | 321 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |