Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chmp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017844.1 | NP_001017844.1 | 1 - 2 | 330 | 79 | |
GenBank mRNA | BC092770.1 | - | 1 - 2 | 330 | 79 | |
BC165146.1 | - | 1 - 2 | 276 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |